Learn More
MedChemExpress Tris(4-aminophenyl)methane
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
Specifications
Specifications
| Content And Storage | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Product Type | Dye Reagents |
| Quantity | 500 mg |
| Shipping Condition | Room temperature |
| Synonym | Leucopararosaniline |
| CAS | 548-61-8 |
| Molecular Weight (g/mol) | 289.37 g/mol |
| Solubility | DMSO : 50 mg/mL (172.79 mM; Need ultrasonic) |
| Target | HBV |