missing translation for 'onlineSavingsMsg'
Learn More

Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate, Thermo Scientific Chemicals

Catalog Number p-7049824
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
This item is not returnable. View return policy
16165-32-5
MFCD00150160
Trichlorotris(ethylenediamine)chromium(III)
This item is not returnable. View return policy
16165-32-5
MFCD00150160
Trichlorotris(ethylenediamine)chromium(III)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow to Orange
Linear Formula Cr(H2NCH2CH2NH2)3Cl3·3.5H2O
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 401.71 (338.65 Anhydrous)
Physical Form Crystalline
Chemical Name or Material Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'