missing translation for 'onlineSavingsMsg'
Learn More

TRIS, MP Biomedicals™

Product Code. 11481672 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11481672

Brand: MP Biomedicals 04819620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TRIS Ultra Pure, ≥99.9%, White crystalline powder

  • Tris is used in pH control in vitro and in vivo for body fluids and in buffering systems for electrophoresis applications.
  • Tris is used in assays used to characterize the activity and kinetics of the enzymes that catalyze SUMOylation of Small ubiquitin-like proteins (SUMO) and SUMO-dependent protein-protein interactions.
  • Tris have been useful as buffers in a wide variety of biological systems.
  • It has been used as a starting material for polymers, oxazolones (with carboxylic acids) and oxazolidines (with aldehydes).
  • It does not precipitate calcium salts and is of value in maintaining solubility of manganese salts.
  • It can be used for the direct standardization of a strong acid solution; the equivalence point can be determined either potentiometrically or by use of a suitable indicator such as 3-(4-Dimethylamino-1-naphthylazo)-4-methoxybenzenesulfonic acid.
  • It is RNAse and DNAse-free.Tris is relatively non-hygroscopic;but, if needed, it can be dried at 100°C for up to 4 hours to remove any water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.14
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Tris-[hydroxymethyl]aminomethane, THAM, 2-Amino-2-(hydroxymethyl)- 1,3-propandiol, Tromethamine, Trometamol

Specifications

Melting Point 165°C to 175°C
Boiling Point 219°C to 220°C (at 10 mmHg)
Color White
pH 10 to 11.5 (5% aq soln)
Physical Form Crystalline Powder
Quantity 500 g
Loss on Drying ≤1%
Beilstein 741883
Solubility Information Soluble in water (550mg/mL), ethyleneglycol (79.1mg/mL), methanol (26mg/mL), anhydrous ethanol (14.6mg/mL), 95% ethanol (22.0mg/mL), DMF (14mg/mL), acetone (2mg/mL), ethyl acetate (0.5mg/mL), olive oil (0.4mg/mL), and chloroform (0.05mg/mL)
Grade ≥99.9%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.