missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(trimethylsilyl)silane), TRC
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.5g
Specifications
Specifications
| Chemical Name or Material | Tris(trimethylsilyl)silane |
| CAS | 1873-77-4 |
| Molecular Formula | C9H28Si4 |
| Quantity | 2.5 g |
| Synonym | 1,1,1,3,3,3-Hexamethyl-2-(trimethylsilyl)trisilane, TTMSS, Tris(trimethylsilyl)silane |
| InChI Formula | InChI=1S/C9H28Si4/c1-11(2,3)10(12(4,5)6)13(7,8)9/h10H,1-9H3 |
| SMILES | C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C |
| IUPAC Name | bis(trimethylsilyl)silyl-trimethylsilane |
| Molecular Weight (g/mol) | 248.66 |
| Formula Weight | 248.1268 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Dela en innehållskorrigering