Learn More
Macherey-Nagel™ Tritest™ L pH Paper Roll Refill
Product Code. 10663141 Shop All Macherey-Nagel Products
Click to view available options
pH Range:
1 to 11
Unit Size:
Pack of 3
Product Code. 10663141
Brand: Macherey-Nagel™ 90511
Only null left
Add to basket
Add to basket
Featuring three indicator zones for the highest accuracy readings. Macherey Nagel™ Tritest™ L pH Paper Roll Refills give the user the most precise readings possible in pH measurement.
  • Provides rapid and reliable results
  • Shows three different colors for each full pH unit
  • Specially equipped with a hydrophobic barrier between different indicators
Specifications
pH Range 1 to 11
Format Roll
Quantity 3 rolls
Type pH Paper