missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Triton™ X-100, scintillation grade

Product Code. 10269510 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10269510 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10269510 Supplier Thermo Scientific Chemicals Supplier No. 422355000
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 9002-93-1 | C16H26O2 | 250.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For fluorescence/luminescence spectroscopy

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

TRUSTED_SUSTAINABILITY

Specifications

Color Undesignated
CAS 9002-93-1
Physical Form Liquid
Cloud Point 63° to 69°C (1% in aq. soln.)
Density 1.0670g/mL
Quantity 500 mL
Infrared Spectrum Authentic
Packaging EcoSafPak™
Refractive Index 1.4900 to 1.4920 (20°C, 589nm)
Molecular Formula C16H26O2
MDL Number MFCD00132505
Merck Index 15, 6850
Synonym Polyethylene glycol mono [4-(1,1,3,3-tetramethylbutyl)phenyl] ether
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
Molecular Weight (g/mol) 250.38
PubChem CID 5590
Formula Weight 646.85
Chemical Name or Material Triton™ X-100
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.