Learn More
Invitrogen™ TRK fused gene Recombinant Rabbit Monoclonal Antibody (24GB2115)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557801
 View more versions of this product
Product Code. 30275199
  • € 198.00 / 20µL
Description
TRK fused gene Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TFG protein, a recently identified novel protein, is a human homolog of mouse TFG protein. It contains an N-terminal coiled-coil structure, a SPYGQ-rich region followed by octicosapeptide repeats. Research shows that TFG can modulate SHP-1 activity and is a novel member of the NF-kB pathway. The physiological functions of this protein remain largely elusive, but TFG is thought to function by associating with other proteins, including TANK and NEMO. TFG was first identified as a partner of NTRK1 and is involved in generating the thyroid TRK-T3 oncogene, as well as oncogenic rearrangements with ALK in anaplastic lymphoma and NOR1 in mixoid chondrosarcoma.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRK fused gene
Recombinant Monoclonal
Unconjugated
TFG
AI173908; HMSNP; protein TFG; SPG57; TF6; TFG; Trk-fused gene; TRK-fused gene protein; TRKT3; TRKT3 oncogene
Rabbit
Affinity chromatography
RUO
10342, 21787, 360709
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2115
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q92734
TFG
A synthesized peptide derived from human TRK fused gene (aa 362-400).