Learn More
Invitrogen™ TRK fused gene Recombinant Rabbit Monoclonal Antibody (24GB2115)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557800
This item is not returnable.
View return policy
Description
TRK fused gene Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TFG protein, a recently identified novel protein, is a human homolog of mouse TFG protein. It contains an N-terminal coiled-coil structure, a SPYGQ-rich region followed by octicosapeptide repeats. Research shows that TFG can modulate SHP-1 activity and is a novel member of the NF-kB pathway. The physiological functions of this protein remain largely elusive, but TFG is thought to function by associating with other proteins, including TANK and NEMO. TFG was first identified as a partner of NTRK1 and is involved in generating the thyroid TRK-T3 oncogene, as well as oncogenic rearrangements with ALK in anaplastic lymphoma and NOR1 in mixoid chondrosarcoma.
Specifications
| TRK fused gene | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TFG | |
| AI173908; HMSNP; protein TFG; SPG57; TF6; TFG; Trk-fused gene; TRK-fused gene protein; TRKT3; TRKT3 oncogene | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10342, 21787, 360709 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2115 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q92734 | |
| TFG | |
| A synthesized peptide derived from human TRK fused gene (aa 362-400). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |