missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkA Antibody (994307), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1752A
This item is not returnable.
View return policy
Description
TrkA Monoclonal antibody specifically detects TrkA in Human samples. It is validated for Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF
Specifications
| TrkA | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF | |
| 994307 | |
| Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF-ready | |
| DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A | |
| Mouse myeloma cell line NS0-derived recombinant human TrkA, Ala33-Glu407, Accession # P04629 | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Immunology, Neuroscience, Protein Kinase | |
| 4914 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction