missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkA Antibody (MNAC13), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-81074CL1
This item is not returnable.
View return policy
Description
TrkA Monoclonal antibody specifically detects TrkA in Human samples. It is validated for ELISA,Immunohistochemistry,Surface Plasmon Resonance,Neutralization,Immunocytochemistry/Immunofluorescence
Specifications
| TrkA | |
| Monoclonal | |
| CoraFluor 1 | |
| DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A | |
| Human TrkA ECD. | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Immunology, Neuroscience, Phospho Specific, Protein Kinase | |
| 4914 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| ELISA, Immunohistochemistry, Surface Plasmon Resonance, Neutralization, Immunocytochemistry/Immunofluorescence | |
| MNAC13 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction