missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkA Antibody (OTI5B6), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74623FR
This item is not returnable.
View return policy
Description
TrkA Monoclonal specifically detects TrkA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| TrkA | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| NTRK1 | |
| Recombinant protein fragment corresponding to amino acids 440-796 of human TrkA (NP_002520) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5B6 | |
| Western Blot, Immunohistochemistry | |
| DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A | |
| Mouse | |
| Affinity Purified | |
| Adaptive Immunity, Cancer, Immunology, Neuroscience, Phospho Specific, Protein Kinase | |
| 4914.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction