Learn More
TrkA Antibody (OTI5B6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46266
Product Code. 18740894
  • € 582.00 / 0.10mL
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
TrkA Monoclonal specifically detects TrkA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot 1:2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150
87.3 kDa