Learn More
TrkA Antibody (OTI5B6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46266
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
TrkA Monoclonal specifically detects TrkA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Western Blot 1:2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
| 87.3 kDa |