missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkC Antibody (75213) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3731JF669
This item is not returnable.
View return policy
Description
TrkC Monoclonal antibody specifically detects TrkC in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| TrkC | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C | |
| Mouse myeloma cell line NS0-derived recombinant human TrkC, Cys32-Asp428, Accession # Q16288 | |
| 0.5 mL | |
| Cancer, Phospho Specific, Protein Kinase | |
| 4916 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Western Blot, CyTOF | |
| 75213 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction