missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkC Antibody (OTI2B8), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74626F
This item is not returnable.
View return policy
Description
TrkC Monoclonal specifically detects TrkC in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| TrkC | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| NTRK3 | |
| Full-length protein expressed in 293T cell transfected with human TrkC expression vector. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2B8 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| EC 2.7.10, EC 2.7.10.1, ETS related protein-neurotrophic receptor tyrosine kinase fusion protein, ETV6-NTRK3 fusion, gp145(trkC), GP145-TrkC, Neurotrophic tyrosine kinase receptor type 3, neurotrophic tyrosine kinase, receptor, type 3, NT 3 receptor, NT-3 growth factor receptor, Trk-C, TrkC tyrosine kinase, TRKCneurotrophin 3 receptor, tyrosine kinase receptor C | |
| Mouse | |
| Affinity Purified | |
| Cancer, Phospho Specific, Protein Kinase | |
| 4916.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction