missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRMT10C Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29087-1-AP-20UL
This item is not returnable.
View return policy
Description
The TRMT10C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRMT10C. This antibody recognizes human, mouse, rat antigen. The TRMT10C antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| TRMT10C | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 54931 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1200 μg/mL | |
| FLJ20432; MRPP1 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 40-45 kDa | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu