missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRMT12 Monoclonal Antibody (OTI1C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809537S
This item is not returnable.
View return policy
Description
TRMT12 Monoclonal antibody specifically detects TRMT12 in Human samples. It is validated for Western BlotSpecifications
| TRMT12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95944 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9436 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1C12 | |
| Unconjugated | |
| NCR2 | |
| 4632406N01Rik; AI449275; alpha-amino-alpha-carboxypropyl transferase TYW2; homolog of yeast tRNA methyltransferase; TRM12; Trmt12; tRNA methyltranferase 12; tRNA methyltranferase 12 homolog; tRNA methyltransferase 12; tRNA methyltransferase 12 homolog; tRNA wybutosine-synthesizing protein 2 homolog; tRNA(Phe) (4-demethylwyosine(37)-C(7)) aminocarboxypropyltransferase; tRNA-yW synthesizing protein 2; tRNA-yW-synthesizing protein 2; TYW2 | |
| Human recombinant protein fragment corresponding to amino acids 44-190 of human NCR2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction