Learn More
Invitrogen™ TRMT2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120866
Description
Positive test controls include: Raji, U-87MG, SW480, MCF7, NCI-H460. Immunogen sequence: MAGLKRRVPL HSLRYFISMV GLFSKPGLLP WYARNPPGWS QLFLGTVCKG DFTRVIATKC QKGQKSQKKP SHLGPLDGSW QERLADVVTP LWRLSYEEQL KVKFAAQKKI LQRLESYIQM LNGVSVTTAV PKSERLSCLL HPIIPSPVIN GYRNKSTFSV NRGPDGNPKT VGFYLGTWRD GNVVCVQSNH LKNIPEKHSQ VAQYYEVFLR QSPLEPCLVF HEGGYWRELT VRTNSQGHTM AIITFHPQKL.
Probable S-adenosyl-L-methionine-dependent methyltransferase that catalyzes the formation of 5-methyl-uridine at position 54 (m5U54) in all tRNA. May also have a role in tRNA stabilization or maturation.
Specifications
| TRMT2B | |
| Polyclonal | |
| Unconjugated | |
| TRMT2B | |
| CXorf34; dJ341D10.3; TRM2 homolog; TRM2 tRNA methyltransferase 2 homolog B; TRMT2B; tRNA (uracil(54)-C(5))-methyltransferase homolog; tRNA (uracil-5-)-methyltransferase homolog; tRNA methyltransferase 2 homolog B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 503462, 79979 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.47 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96GJ1 | |
| TRMT2B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human TRMT2B (NP_079193.2). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.