Learn More
Invitrogen™ TRMT61B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596917
Description
Immunogen sequence: MLMAWCRGPV LLCLRQGLGT NSFLHGLGQE PFEGARSLCC RSSPRDLRDG EREHEAAQRK APGAESCPSL PLSISDIGTG CLSSLENLRL PTLREESSPR ELEDSSGDQG RCGPTHQGSE DPSMLSQAQS ATEVEERHVS PSCSTSRERP FQAGELILAE TGEGETKFKK LFRLNNFGLL NSNWGAVPFG KIVGKFPGQI LRSSFGKQYM LRRPALEDYV VLMKRGTAIT FPKDINMILS MMDINPGDTV; Positive Samples: Mouse liver; Cellular Location: Mitochondrion.
Methyltransferase that catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in various tRNAs in mitochondrion, including tRNA(Leu) (decephering codons UUA or UUG), tRNA(Lys) and tRNA(Ser) (decephering codons UCA, UCU, UCG or UCC).
Specifications
| TRMT61B | |
| Polyclonal | |
| Unconjugated | |
| TRMT61B | |
| 1110039F03Rik; 623451; Gm6431; mRNA methyladenosine-N(1)-methyltransferase; potential tRNA (adenine(58)-N(1))-methyltransferase catalytic subunit TRMT61B; potential tRNA (adenine-N(1)-)-methyltransferase catalytic subunit TRMT61B; TRMT61B; tRNA (adenine(58)-N(1))-methyltransferase, mitochondrial; tRNA methyltransferase 61 homolog B; tRNA methyltransferase 61B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55006 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BVS5 | |
| TRMT61B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human TRMT61B (NP_0603803). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.