missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRMT9B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93012-0.02ml
This item is not returnable.
View return policy
Description
TRMT9B Polyclonal antibody specifically detects TRMT9B in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TRMT9B | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| C8orf79, EC 2.1.1.-, KIAA1456 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 110-330 of human KIAA1456 (NP_065895.2). VIHHFSTKQRRIRAIKEMARVLVPGGQLMIYVWAMEQKNRHFEKQDVLVPWNRALCSQLFSESSQSGRKRQCGYPERGHPYHPPCSECSCSVCFKEQCGSKRSHSVGYEPAMARTCFANISKEGEEEYGFYSTLGKSFRSWFFSRSLDESTLRKQIERVRPLKNTEVWASSTVTVQPSRHSSLDFDHQEPFSTKGQSLDEEVFVESSSGKHLEWLRAPGTL | |
| 0.02 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 57604 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction