missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRMU Antibody (OTI2A4), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74629AF405
This item is not returnable.
View return policy
Description
TRMU Monoclonal specifically detects TRMU in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| TRMU | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TRMU | |
| Full length recombinant protein of human TRMU (NP_060476) produced in HEK293T cells. | |
| 0.1 mL | |
| 55687.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI2A4 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| EC 2.8.1, EC 2.8.1.-, FLJ10140, mitochondrial 5-methylaminomethyl-2-thiouridylate-methyltransferase, mitochondrial tRNA-specific 2-thiouridylase 1, MTO2 homolog, MTO2MGC99627, MTU1, TRMT, TRMT1, tRNA (5-methylaminomethyl-2-thiouridylate)-methyltransferase, tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase, TRNT1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction