Learn More
Syngene™ Trolley with Feet
GREENER_CHOICE
Product Code. 16534341 Shop All Syngene Products
Change view
Click to view available options
Width (Metric):
81.28 cm
121.92 cm
Unit Size:
Each
Product Code. Width (Metric) unitSize
16534341 81.28 cm Each
16574341 121.92 cm Each
2 options
Product Code. 16534341
Brand: Syngene™ SYN32TRF
Fixed trolley equipped with 4 feet.
  • 4 leveling feet
Specifications
Width (English) 32 in.
Includes Feet
Width (Metric) 81.28 cm
Product Type Trolley