missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROP-2 Antibody (046), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90742AF405
This item is not returnable.
View return policy
Description
TROP-2 Monoclonal specifically detects TROP-2 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TROP-2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Has cross-reactivity in ELISA with: Human TROP2 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 46 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TROP-2 (Q8BGV3; Met1-Gln270). | |
| 0.1 mL | |
| Signal Transduction | |
| 4070 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction