missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROP-2 Antibody (MM0588-49D6), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11982PEATT594
This item is not returnable.
View return policy
Description
TROP-2 Monoclonal specifically detects TROP-2 in Human samples. It is validated for Flow Cytometry.
Specifications
| TROP-2 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| TACSTD2 | |
| Human recombinant TROP-2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2 |
| Flow Cytometry | |
| MM0588-49D6 | |
| Flow Cytometry | |
| Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2 | |
| Mouse | |
| Protein G purified | |
| Signal Transduction | |
| 4070.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction