missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROP-2 Rabbit anti-Human, Clone: 20, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 379.00 - € 552.00
Specifications
| Antigen | TROP-2 |
|---|---|
| Clone | 20 |
| Dilution | Immunocytochemistry/Immunofluorescence 1:20-1:100 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18230487
|
Novus Biologicals
NBP2-89493-50UL |
50 μg |
€ 379.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18226918
|
Novus Biologicals
NBP2-89493-100UL |
100 μg |
€ 552.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TROP-2 Monoclonal specifically detects TROP-2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| TROP-2 | |
| Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2 | |
| Recombinant Human TROP-2 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 20 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 4070 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title