Learn More
Invitrogen™ TROP2 Recombinant Rabbit Monoclonal Antibody (064)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA529826
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse TROP-2/TACSTD2.
TROP2 is a carcinoma-associated antigen defined by the monoclonal antibody GA733. This antigen is a member of a family including at least two type I membrane proteins. It transduces an intracellular calcium signal and acts as a cell surface receptor. Mutations of its gene result in gelatinous drop-like corneal dystrophy, an autosomal recessive disorder characterized by severe corneal amyloidosis leading to blindness.
Specifications
| TROP2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q8BGV3 | |
| TACSTD2 | |
| Recombinant Mouse TROP-2/TACSTD2 protein (Met1-Gly270). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 064 | |
| Unconjugated | |
| TACSTD2 | |
| 40kD glycoprotein, identified by monoclonal antibody GA733; C80403; cell surface glycoprotein TROP2; cell surface glycoprotein Trop-2; EGP1; EGP-1; epithelial glycoprotein-1; GA733; GA7331; GA733-1; gastrointestinal tumor-associated antigen GA7331; GP50; Ly97; lymphocyte antigen 97; M1S1; Membrane component chromosome 1 surface marker 1; membrane component, chromosome 1, surface marker 1; pancreatic carcinoma marker protein GA7331; pancreatic carcinoma marker protein GA733-1; parturition-related protein 1; Prp1; TACD2; TACSTD2; Trop2; truncated TACSTD2; tumor-associated calcium signal transducer 2; tunor-associated calcium signal transducer 2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 56753 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.