missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Trophinin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119479
This item is not returnable.
View return policy
Description
Trophinin Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a membrane protein that mediates cell adhesion between trophoblastic cells and the epithelial cells of the endometrium. The encoded protein participates in cell signalling during embryo implantation, and may also be involved in cancer formation. This gene is located near several other closely related genes on chromosome X. Alternative splicing results in multiple transcript variants.
Specifications
| Trophinin | |
| Polyclonal | |
| Unconjugated | |
| TRO | |
| AA409408; KIAA1114; LOC100912141; MAGE superfamily protein; MAGED3; MAGE-d3; MAGE-D3 antigen; Maged3l; magphinin; melanoma antigen, family D, 3; necdin and trophinin like; Tnn; TRO; Trol; trophinin; trophinin-2; trophinin-like | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 7216 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q12816 | |
| TRO | |
| Recombinant protein Trophinin. The antigen corresponds to amino acid range 444-642 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction