Learn More
Tropifexor, MedChemExpress
Product Code. 16481079
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481079 1 mg 1mg
16436827 5 mg 5mg
16435096 100 mg 100mg
16434128 10 mg 10mg
16400487 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16481079 Supplier Medchem Express Supplier No. HY107418
MedChemExpress Tropifexor (LJN452) is a highly potent agonist of FXR with an EC50 of 0.2 nM.
Tropifexor (LJN452) is a highly potent agonist of FXR with an EC50 of 0.2 nM.
Specifications
Chemical Name or Material Tropifexor
CAS 1383816-29-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H25F4N3O5S
Quantity 1 mg
Synonym LJN452
Solubility Information DMSO : ≥ 80.66 mg/mL (133.64 mM)
SMILES FC(F)(F)OC(C=CC=C1)=C1C2=NOC(C3CC3)=C2CO[C@@H]4C[C@H]5N(C6=NC7=C(F)C=C(C(O)=O)C=C7S6)[C@H](CC5)C4
Molecular Weight (g/mol) 603.58
Formula Weight 603.58
Percent Purity 98.34%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only