missing translation for 'onlineSavingsMsg'
Learn More

Tropifexor, MedChemExpress

Product Code. 16435096
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16435096 100 mg 100mg
16481079 1 mg 1mg
16436827 5 mg 5mg
16434128 10 mg 10mg
16400487 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16435096 Supplier Medchem Express Supplier No. HY107418

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tropifexor (LJN452) is a highly potent agonist of FXR with an EC50 of 0.2 nM.

Tropifexor (LJN452) is a highly potent agonist of FXR with an EC50 of 0.2 nM.

Specifications

Chemical Name or Material Tropifexor
CAS 1383816-29-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H25F4N3O5S
Quantity 100 mg
Synonym LJN452
Solubility Information DMSO : ≥ 80.66 mg/mL (133.64 mM)
SMILES FC(F)(F)OC(C=CC=C1)=C1C2=NOC(C3CC3)=C2CO[C@@H]4C[C@H]5N(C6=NC7=C(F)C=C(C(O)=O)C=C7S6)[C@H](CC5)C4
Molecular Weight (g/mol) 603.58
Formula Weight 603.58
Percent Purity 98.34%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.