missing translation for 'onlineSavingsMsg'
Learn More

Tropisetron, MedChemExpress

Product Code. 16458076
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16458076

Brand: Medchem Express HYB0072

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tropisetron (SDZ-ICS-930 free base) is a selective 5-HT3 receptor antagonist and α7-nicotinic receptor agonist with an IC50 of 70.1 ± 0.9 nM for 5-HT3 receptor.

Tropisetron (SDZ-ICS-930 free base) is a selective 5-HT3 receptor antagonist and α7-nicotinic receptor agonist with an IC50 of 70.1 ± 0.9 nM for 5-HT3 receptor.

Specifications

Chemical Name or Material Tropisetron
Physical Form Solid
CAS 89565-68-4
Color Orange
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H20N2O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym SDZ-ICS-930 free base
Solubility Information DMSO : ≥ 100 mg/mL (351.68 mM) ∣H2O : ≥ 55 mg/mL (193.42 mM)
SMILES O=C(C1=CNC2=C1C=CC=C2)O[C@H]3C[C@H]4CC[C@H](N4C)C3
Molecular Weight (g/mol) 284.35
Formula Weight 284.35
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.