missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tropomodulin 1 Antibody (OTI2C2), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74632JF549
This item is not returnable.
View return policy
Description
Tropomodulin 1 Monoclonal specifically detects Tropomodulin 1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Tropomodulin 1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TMOD1 | |
| Full length recombinant protein of human Tropomodulin 1 (NP_003266) produced in HEK293T cells. | |
| 0.1 mL | |
| 7111.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2C2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| D9S57E, Erythrocyte tropomodulin, E-Tmod, e-tropomodulin, TMODETMOD, tropomodulin 1, tropomodulin-1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction