missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tropomodulin 4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | Tropomodulin 4 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18608152
|
Novus Biologicals
NBP2-95207-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653272
|
Novus Biologicals
NBP2-95207-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tropomodulin 4 Polyclonal antibody specifically detects Tropomodulin 4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Tropomodulin 4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 29765 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| actin-capping protein, DKFZp779I0852, Skeletal muscle tropomodulin, SK-TMOD, tropomodulin 4 (muscle), tropomodulin-4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-345 of human TMOD4 (NP_037485.2). MSSYQKELEKYRDIDEDEILRTLSPEELEQLDCELQEMDPENMLLPAGLRQRDQTKKSPTGPLDREALLQYLEQQALEVKERDDLVPFTGEKKGKPYIQPKREIPAEEQITLEPELEEALAHATDAEMCDIAAILDMYTLMSNKQYYDALCSGEICNTEGISSVVQPDKYKPVPDEPPNPTNIEEILKRVRSNDKELEEVNLNNIQDIPIPMLSELCEAMKANTYVRSFSLVATRSGDPIANAVADMLRENRSLQSLNIESNFISSTGLMAVLKAVRENATLTELRVDNQRQWPGDAVEMEMATVLEQCPSIVRFGYHFTQQGPRARAAQAMTRNNELRRQQKKR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title