missing translation for 'onlineSavingsMsg'
Learn More

Troponin C (cardiac) Antibody, CoraFluor™ 1, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP2-99393CL1

Product Code. 30145363

  • € 822.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Troponin C (cardiac) Polyclonal antibody specifically detects Troponin C (cardiac) in Cynomolgus Monkey,Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Troponin C (cardiac)
Polyclonal
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Cynomolgus Monkey, Human
Purified
Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CoraFluor 1
cardiac troponin C, CMD1Z, CMH13, slow twitch skeletal/cardiac muscle troponin C, TNC, TN-C, TNNC, troponin C type 1 (slow), troponin C, slow, troponin C, slow skeletal and cardiac muscles, troponin C1, slow
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Troponin C (cardiac).
0.1 mL
Cell Biology, Cellular Markers
7134
Store at 4°C in the dark. Do not freeze.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.