missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troponin C (cardiac) Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97146FR
This item is not returnable.
View return policy
Description
Troponin C (cardiac) Polyclonal specifically detects Troponin C (cardiac) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Troponin C (cardiac) | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| cardiac troponin C, CMD1Z, CMH13, slow twitch skeletal/cardiac muscle troponin C, TNC, TN-C, TNNC, troponin C type 1 (slow), troponin C, slow, troponin C, slow skeletal and cardiac muscles, troponin C1, slow | |
| Produced in rabbits immunized with E. coli-derived Human Troponin C (cardiac) fragment. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 7134 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction