missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troponin C (cardiac) Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99435-100ul
This item is not returnable.
View return policy
Description
Troponin C (cardiac) Polyclonal antibody specifically detects Troponin C (cardiac) in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Troponin C (cardiac) | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| cardiac troponin C, CMD1Z, CMH13, slow twitch skeletal/cardiac muscle troponin C, TNC, TN-C, TNNC, troponin C type 1 (slow), troponin C, slow, troponin C, slow skeletal and cardiac muscles, troponin C1, slow | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Troponin C (cardiac) | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 7134 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction