missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troponin C (cardiac) Antibody (SR1283), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21985-50ul
This item is not returnable.
View return policy
Description
Troponin C (cardiac) Monoclonal antibody specifically detects Troponin C (cardiac) in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Troponin C (cardiac) | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1283 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| cardiac troponin C, CMD1Z, CMH13, slow twitch skeletal/cardiac muscle troponin C, TNC, TN-C, TNNC, troponin C type 1 (slow), troponin C, slow, troponin C, slow skeletal and cardiac muscles, troponin C1, slow | |
| A synthesized peptide derived from human Troponin C (cardiac) (Uniprot #: P63316) | |
| 50 μg | |
| Cell Biology, Cellular Markers | |
| 7134 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction