Learn More
Invitrogen™ Troponin I Recombinant Superclonal™ Antibody (1HCLC)
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 710580
Description
Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.
Troponin proteins associate with tropomyosin and regulate the calcium sensitivity of the myofibril contractile apparatus of striated muscles. Troponin I (TnI), along with troponin T (TnT) and troponin C (TnC), is one of 3 subunits that form the troponin complex of the thin filaments of striated muscle. TnI is the inhibitory subunit; blocking actin-myosin interactions and thereby mediating striated muscle relaxation. The TnI subfamily contains three genes: TnI-skeletal-fast-twitch, TnI-skeletal-slow-twitch, and TnI-cardiac. The TnI-fast and TnI-slow genes are expressed in fast-twitch and slow-twitch skeletal muscle fibers, respectively, while the TnI-cardiac gene is expressed exclusively in cardiac muscle tissue. This gene encodes the Troponin-I-skeletal-slow-twitch protein. This gene is expressed in cardiac and skeletal muscle during early development but is restricted to slow-twitch skeletal muscle fibers in adults. The encoded protein prevents muscle contraction by inhibiting calcium-mediated conformational changes in actin-myosin complexes.
Specifications
| Troponin I | |
| Recombinant Superclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P13413, P19237, P19429, P23693, P48787, Q9WUZ5 | |
| TNNI1, TNNI3 | |
| human Troponin I. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1HCLC | |
| Unconjugated | |
| TNNI1 | |
| 2700018B22Rik; AI747285; Cardiac troponin I; cardiomyopathy, dilated 2A (autosomal recessive); CMD1FF; CMD2A; CMH7; cTNI; DKFZp451O223; RCM1; slow skeletal troponin I isoform; ssTnI; Tn1; TnI; TNN1; TNNC1; TNNI1; TNNI3; Tpn1; troponin 1, type 3; troponin I type 1 (skeletal, slow); troponin I type 3 (cardiac); troponin I, cardiac 3; troponin I, cardiac muscle; troponin I, skeletal, slow; troponin I, skeletal, slow 1; Troponin I, slow isoform; troponin I, slow skeletal; troponin I, slow skeletal muscle; Troponin I, slow-twitch isoform; troponin I1, slow skeletal type; troponin I3, cardiac type | |
| Rabbit | |
| Protein A | |
| RUO | |
| 21952, 21954, 29248, 29388, 7135, 7137 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.