missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TROVE2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521454
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HepG2, Molt-4, Raji, mouse heart. Predicted reactivity: Mouse (89%), Rat (88%), Xenopus laevis (80%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TROVE2 belongs to the Ro 60 kDa family. It is RNA-binding protein that binds to several small cytoplasmic RNA molecules known as Y RNAs. It may stabilize these RNAs from degradation.
Specifications
| TROVE2 | |
| Polyclonal | |
| Unconjugated | |
| RO60 | |
| 1810007I17Rik; 60 kDa ribonucleoprotein Ro; 60 kDa Ro protein; 60 kDa SS-A/Ro ribonucleoprotein; 60 kDa SS-A/Ro ribonucleoprotein-like protein; A530054J02Rik; AI646302; gastric cancer multi-drug resistance protein; LOW QUALITY PROTEIN: 60 kDa SS-A/Ro ribonucleoprotein; Ro 60 kDa autoantigen; RO60; Ro60 autoantigen; Ro60, Y RNA binding protein; roRNP; RP11-101E13.3; Sjoegren syndrome antigen A2; Sjoegren syndrome type A antigen; Sjogren syndrome antigen A2; Sjogren syndrome antigen A2 (60kD, ribonucleoprotein autoantigen SS-A/Ro); Ssa; SS-A; SS-A/Ro; SSA2; TROVE domain family member 2; TROVE domain family, member 2; Trove2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20822, 6738 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O08848, P10155 | |
| RO60 | |
| Recombinant protein encompassing a sequence within the center region of human TROVE2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction