missing translation for 'onlineSavingsMsg'
Learn More

Troxipide, MedChemExpress

Product Code. 16458446
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16458446

Brand: Medchem Express HYB0758

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Troxipide is an orally active defensive factor-enhancing therapeutic agent for gastritis and gastric ulcer (GU). Troxipide is a non-antisecretory gastro protective agent with antiulcer, anti-inflammatory and mucus-secreting properties.

Troxipide is an orally active defensive factor-enhancing therapeutic agent for gastritis and gastric ulcer (GU). Troxipide is a non-antisecretory gastro protective agent with antiulcer, anti-inflammatory and mucus-secreting properties.

Specifications

Chemical Name or Material Troxipide
CAS 30751-05-4
Color White
Purity Grade Notes Research
Molecular Formula C15H22N2O4
Quantity 100 mg
Solubility Information DMSO : 9.09 mg/mL (30.88 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.40 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(NC1CNCCC1)C2=CC(OC)=C(OC)C(OC)=C2
Molecular Weight (g/mol) 294.35
Formula Weight 294.35
Percent Purity 99.45%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.