Learn More
TROY/TNFRSF19 Antibody (933202) [DyLight 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB15481E
Product Code. 18563756
  • ONLINE EXCLUSIVE PRICE
    € 657.00
    € 620.55 / 0.10mL
    Save € 36.45
    5% Off
Description
TROY/TNFRSF19 Monoclonal antibody specifically detects TROY/TNFRSF19 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TROY/TNFRSF19
Monoclonal
DyLight 405
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
933202
Flow Cytometry, CyTOF-ready
TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19
Mouse myeloma cell line NS0-derived recombinant human TROY/TNFRSF19, Glu30-Leu170, Accession # Q9NS68
0.1 mL
Apoptosis
55504
Store at 4°C in the dark.