missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TROY/TNFRSF19 Antibody (933202), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB15481PECY7
This item is not returnable.
View return policy
Description
TROY/TNFRSF19 Monoclonal antibody specifically detects TROY/TNFRSF19 in Human samples. It is validated for Flow Cytometry
Specifications
| TROY/TNFRSF19 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 933202 | |
| Flow Cytometry | |
| TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
| Mouse myeloma cell line NS0-derived recombinant human TROY/TNFRSF19, Glu30-Leu170, Accession # Q9NS68 | |
| 0.1 mL | |
| Apoptosis | |
| 55504 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction