missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPC3 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32258AF750
This item is not returnable.
View return policy
Description
TRPC3 Polyclonal antibody specifically detects TRPC3 in Human samples. It is validated for Western Blot
Specifications
| TRPC3 | |
| Polyclonal | |
| Western Blot | |
| hTrp3, hTrp-3, short transient receptor potential channel 3, transient receptor potential cation channel, subfamily C, member 3, transient receptor potential channel 3, Transient receptor protein 3, TRP3, TRP-3, TrpC3 | |
| The immunogen this antibody was made to, maps to a region between residue 275 to 325 of human Transient Receptor Potential Cation Channel, Subfamily C, Member 3 using the numbering given in entry Q13507.3 (GeneID 7222). | |
| 0.1 mL | |
| Cardiovascular Biology, Neuroscience, Signal Transduction | |
| 7222 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction