missing translation for 'onlineSavingsMsg'
Learn More

TRPC5 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18615830 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18615830 0.02 mL 0.02mL
18617801 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18615830 Supplier Novus Biologicals Supplier No. NBP2945850.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRPC5 Polyclonal antibody specifically detects TRPC5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPC5
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias hTRP5, hTRP-5, short transient receptor potential channel 5, transient receptor potential cation channel, subfamily C, member 5, Transient receptor protein 5, TRP-5, TRP5transient receptor potential channel 5, TrpC5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 800-900 of human TRPC5 (NP_036603.1). NLGCKKKTCHGPPLIRTMPRSSGAQGKSKAESSSKRSFMGPSLKKLGLLFSKFNGHMSEPSSEPMYTISDGIVQQHCMWQDIRYSQMEKGKAEACSQSEIN
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7224
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.