missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TRPC6 Antibody (TRPC6/7671) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27020H
This item is not returnable.
View return policy
Description
TRPC6 Monoclonal antibody specifically detects TRPC6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TRPC6 | |
| Monoclonal | |
| HRP | |
| FLJ11098, FSGS2, short transient receptor potential channel 6, transient receptor potential cation channel, subfamily C, member 6, Transient receptor protein 6, TRP-6, TRP6FLJ14863, TrpC6 | |
| Recombinant full-length human TRPC6 protein | |
| 0.1 mL | |
| Signal Transduction | |
| 7225 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| TRPC6/7671 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction