missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TrpE Tag Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116407
This item is not returnable.
View return policy
Description
TrpE Tag Polyclonal Antibody for Western Blot
TrpE is a commonly used tag in some protein expression systems. TrpE antibodies can be used for analysis of expression of proteins alone or in conjunction with protein specific antibodies. They can also be used as a negative control, as they do not react with any known mammalian protein.
Specifications
| TrpE Tag | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| KLH conjugated synthetic peptide between391-422 amino acids from the Bifidobacterium TrpE. | |
| 200 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Tag |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| Tag | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction