missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48635-25ul
This item is not returnable.
View return policy
Description
TRPM2 Polyclonal antibody specifically detects TRPM2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TRPM2 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| EC 3.6.1.13, EREG1MGC133383, Estrogen-responsive element-associated gene 1 protein, KNP3LTrpC-2, Long transient receptor potential channel 2, LTrpC2, LTRPC2TRPC7transient receptor potential cation channel subfamily M member 2, NUDT9H, NUDT9L1, transient receptor potential cation channel, subfamily M, member 2, Transient receptor potential channel 7, TrpC7 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RTRLEKFISEQTKERGGVAIKIPIVCVVLEGGPGTLHTIDNATTNGTPCVVVEGSGRVADVIAQVANLPVSDITISLIQQKLSVFFQEMFETFTESRIVEWTKKIQDIVRRRQLLTVFREGKDGQQDV | |
| 25 μL | |
| Apoptosis, Cell Biology, Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
| 7226 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction