Learn More
TRPM2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21842-1-AP-150UL
 View more versions of this product
Product Code. 18065295
  • € 410.00 / 150µL
Description
The TRPM2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TRPM2. This antibody recognizes human, mouse antigen. The TRPM2 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Specifications
TRPM2
1000 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Immunofluorescence, Immunohistochemistry, ELISA
EREG1, KNP3, LTrpC 2, LTRPC2, NUDT9H, NUDT9L1, TRPC7, TRPM2
150 μL
7226
Store at -20°C. Stable for one year after shipment.