missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Progesterone ELISA Kit (Colorimetric)

Product Code. 18065296 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18065296 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18065296 Supplier Novus Biologicals™ Supplier No. NBP2601241KIT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High sensitivity and specificity for detection of human progesterone.

The Human Progesterone ELISA Kit (Colorimetric) measures human Progesterone in serum,plasma.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.4 to 30ng/mL
Assay Sensitivity 0.2ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum
Specificity For detection of human PROG
Target Species Human
For Use With (Application) ELISA
Kit Contents Instruction manual, Standard, Stop Solution, Wash Buffer (20 x concentrate), Substrate B, Substrate A, HRP-conjugate, Assay plate, Antibody, Adhesive Strip (For 96 wells)
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Signal Transduction
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym Pregn 4 ene 3 20 dione
Target Progesterone
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.