missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM4 Antibody (OTI10H5), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74647F
This item is not returnable.
View return policy
Description
TRPM4 Monoclonal specifically detects TRPM4 in Human, Canine, Primate, Monkey samples. It is validated for Immunohistochemistry.
Specifications
| TRPM4 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| TRPM4 | |
| Full length recombinant protein of human TRPM4 (NP_060106) produced in HEK293T cells. | |
| 0.1 mL | |
| 54795.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI10H5 | |
| Immunohistochemistry | |
| Calcium-activated non-selective cation channel 1, FLJ20041, hTRPM4, Long transient receptor potential channel 4, LTrpC-4, LTRPC4, melastatin-4, PFHB1B, transient receptor potential cation channel subfamily M member 4, transient receptor potential cation channel, subfamily M, member 4, TRPM4B | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Canine, Primate, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction