missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM4 Mouse anti-Canine, Human, Clone: OTI10H5, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500381S
This item is not returnable.
View return policy
Description
Calcium-activated non selective (CAN) cation channel that mediates membrane depolarization. While it is activated by increase in intracellular Ca2+, it is impermeable to it. Mediates transport of monovalent cations (Na+, K+, Cs+, Li+), leading to depolarize the membrane. It thereby plays a central role in cadiomyocytes, neurons from entorhinal cortex, dorsal root and vomeronasal neurons, endocrine pancreas cells, kidney epithelial cells, cochlea hair cells etc. Participates in T-cell activation by modulating Ca2+ oscillations after T lymphocyte activation, which is required for NFAT-dependent IL2 production. Involved in myogenic constriction of cerebral arteries. Controls insulin secretion in pancreatic beta-cells. May also be involved in pacemaking or could cause irregular electrical activity under conditions of Ca2+ overload. Affects T-helper 1 (Th1) and T-helper 2 (Th2) cell motility and cytokine production through differential regulation of calcium signaling and NFATC1 localization. Enhances cell proliferation through up-regulation of the beta-catenin signaling pathway.Specifications
| TRPM4 | |
| Monoclonal | |
| Unconjugated | |
| TRPM4 | |
| LTrpC4, PFHB1B, TRPM4B, hTRPM4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 484385, 54795 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI10H5 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TD43 | |
| TRPM4 | |
| Full length human recombit protein of human TRPM4 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction