missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPM5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94006-0.1ml
This item is not returnable.
View return policy
Description
TRPM5 Polyclonal antibody specifically detects TRPM5 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TRPM5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Long transient receptor potential channel 5, LTrpC5, LTrpC-5, LTRPC5transient receptor potential cation channel subfamily M member 5, MLSN1- and TRP-related gene 1 protein, MTR1MLSN1 and TRP-related, transient receptor potential cation channel, subfamily M, member 5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1026-1165 of human TRPM5 (NP_055370.1). RVFKKEAEHKREHLERDLPDPLDQKVVTWETVQKENFLSKMEKRRRDSEGEVLRKTAHRVDFIAKYLGGLREQEKRIKCLESQINYCSVLVSSVADVLAQGGGPRSSQHCGEGSQLVAADHRGGLDGWEQPGAGQPPSDT | |
| 0.1 mL | |
| Neuroscience | |
| 29850 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction